Product Details

SNP ID
rs201922786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141193317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGACCCAGGCTCCAGAAAACAG[C/T]CCCATTGGGTTCCTTATTGTTAAGG
Phenotype
MIM: 606336 MIM: 606337 MIM: 606345 MIM: 606335
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDHB10 PubMed Links

Gene Details

Gene
PCDHB10
Gene Name
protocadherin beta 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018930.3 939 Silent Mutation AGC,AGT S255S NP_061753.1
Gene
PCDHB11
Gene Name
protocadherin beta 11
There are no transcripts associated with this gene.

Gene
PCDHB16
Gene Name
protocadherin beta 16
There are no transcripts associated with this gene.

Gene
PCDHB9
Gene Name
protocadherin beta 9
There are no transcripts associated with this gene.

View Full Product Details