Product Details

SNP ID
rs200051899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140547817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTACAGCACCACGCCGGGGGGCA[A/C]GCTATACGCCACTACCCCCGGAGGT
Phenotype
MIM: 610500 MIM: 603483 MIM: 603819
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKHD1 PubMed Links

Gene Details

Gene
ANKHD1
Gene Name
ankyrin repeat and KH domain containing 1
There are no transcripts associated with this gene.

Gene
ANKHD1-EIF4EBP3
Gene Name
ANKHD1-EIF4EBP3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020690.5 152 Intron NP_065741.3
Gene
EIF4EBP3
Gene Name
eukaryotic translation initiation factor 4E binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003732.2 152 Missense Mutation AAG,ACG K27T NP_003723.1
Gene
SRA1
Gene Name
steroid receptor RNA activator 1
There are no transcripts associated with this gene.

View Full Product Details