Product Details
- SNP ID
-
rs200230756
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:55626576 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGATTAATTTAGGCCATGTCAGA[C/T]GCTCTTCTTGGTGGAGGGAAATTAT
- Phenotype
-
MIM: 616203
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLC38A9
PubMed Links
Gene Details
- Gene
- SLC38A9
- Gene Name
- solute carrier family 38 member 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001258286.1 |
2027 |
Missense Mutation |
CAT,CGT |
H472R |
NP_001245215.1 |
NM_001258287.1 |
2027 |
Missense Mutation |
CAT,CGT |
H472R |
NP_001245216.1 |
NM_001282429.1 |
2027 |
Missense Mutation |
CAT,CGT |
H411R |
NP_001269358.1 |
NM_173514.3 |
2027 |
Missense Mutation |
CAT,CGT |
H535R |
NP_775785.2 |
XM_006714537.2 |
2027 |
Missense Mutation |
CAT,CGT |
H535R |
XP_006714600.1 |
XM_006714538.3 |
2027 |
Missense Mutation |
CAT,CGT |
H535R |
XP_006714601.1 |
XM_006714539.3 |
2027 |
Missense Mutation |
CAT,CGT |
H535R |
XP_006714602.1 |
XM_011543173.1 |
2027 |
Missense Mutation |
CAT,CGT |
H535R |
XP_011541475.1 |
XM_011543174.1 |
2027 |
Missense Mutation |
CAT,CGT |
H535R |
XP_011541476.1 |
XM_011543176.1 |
2027 |
Missense Mutation |
CAT,CGT |
H505R |
XP_011541478.1 |
XM_011543177.1 |
2027 |
Missense Mutation |
CAT,CGT |
H499R |
XP_011541479.1 |
XM_011543178.1 |
2027 |
Missense Mutation |
CAT,CGT |
H478R |
XP_011541480.1 |
XM_011543180.1 |
2027 |
Missense Mutation |
CAT,CGT |
H434R |
XP_011541482.1 |
XM_011543182.1 |
2027 |
UTR 3 |
|
|
XP_011541484.1 |
XM_011543183.1 |
2027 |
Missense Mutation |
CAT,CGT |
H413R |
XP_011541485.1 |
XM_011543184.1 |
2027 |
Missense Mutation |
CAT,CGT |
H377R |
XP_011541486.1 |
XM_017009077.1 |
2027 |
Missense Mutation |
CAT,CGT |
H505R |
XP_016864566.1 |
XM_017009078.1 |
2027 |
Missense Mutation |
CAT,CGT |
H478R |
XP_016864567.1 |
XM_017009079.1 |
2027 |
Missense Mutation |
CAT,CGT |
H470R |
XP_016864568.1 |
XM_017009080.1 |
2027 |
Missense Mutation |
CAT,CGT |
H470R |
XP_016864569.1 |
XM_017009081.1 |
2027 |
Missense Mutation |
CAT,CGT |
H440R |
XP_016864570.1 |
XM_017009082.1 |
2027 |
Missense Mutation |
CAT,CGT |
H434R |
XP_016864571.1 |
View Full Product Details