Product Details

SNP ID
rs200172929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:123346699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCCATGGGGGCCATCCTTTTT[A/T]CCACTTGCAATCTCTGTGGAATCCT
Phenotype
MIM: 613446
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP120 PubMed Links

Gene Details

Gene
CEP120
Gene Name
centrosomal protein 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166226.1 3231 Silent Mutation GGA,GGT G901G NP_001159698.1
NM_153223.3 3231 Silent Mutation GGA,GGT G927G NP_694955.2
XM_005271901.4 3231 Silent Mutation GGA,GGT G882G XP_005271958.1
XM_011543185.2 3231 Silent Mutation GGA,GGT G901G XP_011541487.1
XM_011543186.2 3231 Silent Mutation GGA,GGT G438G XP_011541488.1
XM_017009085.1 3231 Silent Mutation GGA,GGT G438G XP_016864574.1

View Full Product Details