Product Details

SNP ID
rs199877255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:33936781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCACGATAGCCACCATGAATA[A/C]GGCAGCAGGCGGGGACAAGCTAGCA
Phenotype
MIM: 609445 MIM: 606202
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RXFP3 PubMed Links

Gene Details

Gene
RXFP3
Gene Name
relaxin/insulin like family peptide receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016568.3 396 Missense Mutation AAG,ACG K14T NP_057652.1
Gene
SLC45A2
Gene Name
solute carrier family 45 member 2
There are no transcripts associated with this gene.

View Full Product Details