Product Details

SNP ID
rs200057403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:154950871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGTGCTTCTCTTGACATTTCTC[A/G]AAAATGGGAGAAGAAGAATAAAATT
Phenotype
MIM: 611835
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL22 PubMed Links

Gene Details

Gene
MRPL22
Gene Name
mitochondrial ribosomal protein L22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014990.2 166 Intron NP_001014990.1
NM_014180.3 166 Missense Mutation CAA,CGA Q43R NP_054899.2

View Full Product Details