Product Details

SNP ID
rs200158376
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:58455399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTGAGCATTGGCCAAGCTCTGC[A/G]TAATAGTGAACTGTTCTATAAAAAC
Phenotype
MIM: 607023
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PLK2 PubMed Links

Gene Details

Gene
PLK2
Gene Name
polo like kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252226.1 1821 Silent Mutation TAC,TAT Y533Y NP_001239155.1
NM_006622.3 1821 Silent Mutation TAC,TAT Y547Y NP_006613.2

View Full Product Details