Product Details

SNP ID
rs200140092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132674126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGGCAACTTTGTCCACGGACAC[A/G]AGTGCGATATCACCTTACAGGAGAT
Phenotype
MIM: 147780
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL4 PubMed Links

Gene Details

Gene
IL4
Gene Name
interleukin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000589.3 141 Missense Mutation AAG,GAG K26E NP_000580.1
NM_172348.2 141 Missense Mutation AAG,GAG K26E NP_758858.1
Gene
LOC105379176
Gene Name
uncharacterized LOC105379176
There are no transcripts associated with this gene.

View Full Product Details