Product Details

SNP ID
rs200218141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1878105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGTCAGCACGTTCGCGCCCGCC[C/T]CCAGCGAGCGTCCCAGAGGCCCGGG
Phenotype
MIM: 606199
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTD-2194D22.4 PubMed Links

Gene Details

Gene
CTD-2194D22.4
Gene Name
uncharacterized LOC101929081
There are no transcripts associated with this gene.

Gene
IRX4
Gene Name
iroquois homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278632.1 1642 Missense Mutation GAG,GGG E475G NP_001265561.1
NM_001278633.1 1642 Missense Mutation GAG,GGG E501G NP_001265562.1
NM_001278634.1 1642 Missense Mutation GAG,GGG E475G NP_001265563.1
NM_001278635.1 1642 Missense Mutation GAG,GGG E501G NP_001265564.1
NM_016358.2 1642 Missense Mutation GAG,GGG E475G NP_057442.1

View Full Product Details