Product Details

SNP ID
rs199644520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69419738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGATGGAGTGGAGTGTTCACCAC[C/G]AGCCTCTCCAGCAAGACCAAACCAC
Phenotype
MIM: 610572 MIM: 603139
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MARVELD2 PubMed Links

Gene Details

Gene
MARVELD2
Gene Name
MARVEL domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038603.2 515 Missense Mutation CCA,CGA P118R NP_001033692.2
NM_001244734.1 515 Missense Mutation CCA,CGA P118R NP_001231663.1
XM_005248445.4 515 Missense Mutation CCA,CGA P118R XP_005248502.1
XM_005248446.4 515 Missense Mutation CCA,CGA P118R XP_005248503.1
XM_005248447.4 515 Missense Mutation CCA,CGA P118R XP_005248504.1
Gene
RAD17
Gene Name
RAD17 checkpoint clamp loader component
There are no transcripts associated with this gene.

View Full Product Details