Product Details

SNP ID
rs199684712
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:96393134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTCAGAGTCTTTAAGCTGGGAA[C/G]GTTTGTTCAGCTTTTCCAGGGCTTC
Phenotype
MIM: 162150
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
LOC101929710 PubMed Links

Gene Details

Gene
LOC101929710
Gene Name
uncharacterized LOC101929710
There are no transcripts associated with this gene.

Gene
PCSK1
Gene Name
proprotein convertase subtilisin/kexin type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000439.4 2368 Missense Mutation CCT,CGT P710R NP_000430.3
NM_001177875.1 2368 Missense Mutation CCT,CGT P663R NP_001171346.1

View Full Product Details