Product Details

SNP ID
rs199844766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:80650056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGTCAATTTCACTGGTTGAAA[A/G]TAAGAGACAGCTGAACCCTCGTGGA
Phenotype
MIM: 126060 MIM: 600887
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHFR PubMed Links

Gene Details

Gene
DHFR
Gene Name
dihydrofolate reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000791.3 Intron NP_000782.1
NM_001290354.1 Intron NP_001277283.1
NM_001290357.1 Intron NP_001277286.1
Gene
MSH3
Gene Name
mutS homolog 3
There are no transcripts associated with this gene.

Gene
MTRNR2L2
Gene Name
MT-RNR2-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190470.1 Intron NP_001177399.1

View Full Product Details