Product Details

SNP ID
rs200324612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141638246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCATGTCGGAACCACAGCCTGA[C/T]CTGGAACCGCCCCAACATGGGCTAT
Phenotype
MIM: 605166 MIM: 611213
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCHSD1 PubMed Links

Gene Details

Gene
FCHSD1
Gene Name
FCH and double SH3 domains 1
There are no transcripts associated with this gene.

Gene
HDAC3
Gene Name
histone deacetylase 3
There are no transcripts associated with this gene.

Gene
RELL2
Gene Name
RELT like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130029.1 889 Silent Mutation GAC,GAT D7D NP_001123501.1
NM_173828.4 889 Silent Mutation GAC,GAT D7D NP_776189.3
XM_005268414.3 889 Silent Mutation GAC,GAT D7D XP_005268471.1
XM_011537624.1 889 Silent Mutation GAC,GAT D7D XP_011535926.1
XM_011537625.2 889 Silent Mutation GAC,GAT D7D XP_011535927.1

View Full Product Details