Product Details

SNP ID
rs199745108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178120196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCAGTGCCGCATTCGCCCCTCA[A/G]TGTTCAAGCCTACGGCGGGCAACGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
N4BP3 PubMed Links

Gene Details

Gene
N4BP3
Gene Name
NEDD4 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015111.1 596 Missense Mutation ATG,GTG M117V NP_055926.1
XM_006714834.3 596 Missense Mutation ATG,GTG M117V XP_006714897.1
XM_011534473.1 596 Missense Mutation ATG,GTG M117V XP_011532775.1
XM_011534474.1 596 Missense Mutation ATG,GTG M117V XP_011532776.1

View Full Product Details