Product Details

SNP ID
rs200201511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140654534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTAGGTACGAGCTGAGATTGCC[A/T]GCAAAGAGAAAGAGGAAGAGGAACT
Phenotype
MIM: 600549 MIM: 602137
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IK PubMed Links

Gene Details

Gene
IK
Gene Name
IK cytokine, down-regulator of HLA II
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006083.3 648 Missense Mutation AGC,TGC S180C NP_006074.2
Gene
MIR3655
Gene Name
microRNA 3655
There are no transcripts associated with this gene.

Gene
NDUFA2
Gene Name
NADH:ubiquinone oxidoreductase subunit A2
There are no transcripts associated with this gene.

Gene
TMCO6
Gene Name
transmembrane and coiled-coil domains 6
There are no transcripts associated with this gene.

View Full Product Details