Product Details

SNP ID
rs199669417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149860964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTTTCCCCCCGGCTGATTTCC[C/T]GCGGCTGCTGCAATGAAACAGGAAA
Phenotype
MIM: 180071 MIM: 608886
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE6A PubMed Links

Gene Details

Gene
PDE6A
Gene Name
phosphodiesterase 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000440.2 1703 Silent Mutation GCA,GCG A838A NP_000431.2
XM_011537650.2 1703 Silent Mutation GCA,GCG A543A XP_011535952.1
XM_011537651.2 1703 Silent Mutation GCA,GCG A489A XP_011535953.1
XM_011537652.1 1703 Silent Mutation GCA,GCG A479A XP_011535954.1
XM_011537653.2 1703 Silent Mutation GCA,GCG A479A XP_011535955.1
XM_011537654.2 1703 Silent Mutation GCA,GCG A479A XP_011535956.1
XM_017009572.1 1703 Silent Mutation GCA,GCG A757A XP_016865061.1
Gene
PPARGC1B
Gene Name
PPARG coactivator 1 beta
There are no transcripts associated with this gene.

View Full Product Details