Product Details

SNP ID
rs199670821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:173146931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGAATACTAAAGTAAAAGAGAA[A/G]TTTCAACAGTTGCGTCACAGAATAC
Phenotype
MIM: 603291
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BNIP1 PubMed Links

Gene Details

Gene
BNIP1
Gene Name
BCL2 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205.2 254 Silent Mutation AAA,AAG K50K NP_001196.2
NM_013978.2 254 Silent Mutation AAA,AAG K50K NP_053581.2
NM_013979.2 254 Silent Mutation AAA,AAG K50K NP_053582.2
NM_013980.2 254 Silent Mutation AAA,AAG K50K NP_053583.2
XM_011534638.1 254 Silent Mutation AAA,AAG K50K XP_011532940.1
XM_011534639.1 254 Silent Mutation AAA,AAG K50K XP_011532941.1
Gene
CREBRF
Gene Name
CREB3 regulatory factor
There are no transcripts associated with this gene.

View Full Product Details