Product Details

SNP ID
rs200008043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:23521083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAAAGAATTGTCAAGAACAGCA[A/C]ATTTACTGAATGCAAGTGGCTCAGA
Phenotype
MIM: 609760
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRDM9 PubMed Links

Gene Details

Gene
PRDM9
Gene Name
PR domain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310214.1 600 Intron NP_001297143.1
NM_020227.3 600 Missense Mutation AAT,CAT N138H NP_064612.2

View Full Product Details