Product Details
- SNP ID
-
rs200186614
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:88196485 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTGGATAAATACCCCAGGAATGA[A/T]TACCTAGAAAATCAAAGACACAAAT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM161B
PubMed Links
Gene Details
- Gene
- TMEM161B
- Gene Name
- transmembrane protein 161B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001289007.1 |
1296 |
Missense Mutation |
AAT,ATT |
N397I |
NP_001275936.1 |
NM_001289008.1 |
1296 |
Missense Mutation |
AAT,ATT |
N386I |
NP_001275937.1 |
NM_153354.4 |
1296 |
Intron |
|
|
NP_699185.1 |
XM_006714553.1 |
1296 |
Intron |
|
|
XP_006714616.1 |
XM_006714554.3 |
1296 |
Intron |
|
|
XP_006714617.1 |
XM_006714555.3 |
1296 |
Intron |
|
|
XP_006714618.1 |
XM_006714556.3 |
1296 |
Missense Mutation |
AAT,ATT |
N270I |
XP_006714619.2 |
XM_011543201.2 |
1296 |
Intron |
|
|
XP_011541503.1 |
XM_011543202.2 |
1296 |
Missense Mutation |
AAT,ATT |
N270I |
XP_011541504.2 |
XM_011543203.2 |
1296 |
Missense Mutation |
AAT,ATT |
N252I |
XP_011541505.2 |
XM_011543204.2 |
1296 |
Missense Mutation |
AAT,ATT |
N215I |
XP_011541506.2 |
XM_017009093.1 |
1296 |
Missense Mutation |
AAT,ATT |
N397I |
XP_016864582.1 |
XM_017009094.1 |
1296 |
Missense Mutation |
AAT,ATT |
N386I |
XP_016864583.1 |
XM_017009095.1 |
1296 |
Missense Mutation |
AAT,ATT |
N397I |
XP_016864584.1 |
XM_017009096.1 |
1296 |
Missense Mutation |
AAT,ATT |
N270I |
XP_016864585.1 |
XM_017009097.1 |
1296 |
Missense Mutation |
AAT,ATT |
N270I |
XP_016864586.1 |
XM_017009098.1 |
1296 |
Missense Mutation |
AAT,ATT |
N270I |
XP_016864587.1 |
XM_017009099.1 |
1296 |
Missense Mutation |
AAT,ATT |
N270I |
XP_016864588.1 |
XM_017009100.1 |
1296 |
Missense Mutation |
ATC,TTC |
I354F |
XP_016864589.1 |
XM_017009101.1 |
1296 |
Missense Mutation |
AAT,ATT |
N215I |
XP_016864590.1 |
XM_017009102.1 |
1296 |
Missense Mutation |
AAT,ATT |
N215I |
XP_016864591.1 |
XM_017009103.1 |
1296 |
Intron |
|
|
XP_016864592.1 |
XM_017009104.1 |
1296 |
Missense Mutation |
AAT,ATT |
N118I |
XP_016864593.1 |
XM_017009105.1 |
1296 |
Intron |
|
|
XP_016864594.1 |
XM_017009106.1 |
1296 |
Intron |
|
|
XP_016864595.1 |
XM_017009107.1 |
1296 |
Intron |
|
|
XP_016864596.1 |
XM_017009108.1 |
1296 |
Intron |
|
|
XP_016864597.1 |
XM_017009109.1 |
1296 |
Missense Mutation |
ATC,TTC |
I238F |
XP_016864598.1 |
XM_017009110.1 |
1296 |
Missense Mutation |
ATC,TTC |
I238F |
XP_016864599.1 |
View Full Product Details