Product Details

SNP ID
rs201258702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115126783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAATTTCTGCATGCCTATAGTA[A/G]CTAAGGTAAATGGTTGTGAAGAATC
Phenotype
MIM: 609317
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM36 PubMed Links

Gene Details

Gene
TRIM36
Gene Name
tripartite motif containing 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017397.1 2428 Intron NP_001017397.1
NM_001017398.1 2428 Intron NP_001017398.1
NM_001300752.1 2428 Missense Mutation GCT,GTT A481V NP_001287681.1
NM_001300759.1 2428 Missense Mutation GCT,GTT A624V NP_001287688.1
NM_018700.3 2428 Missense Mutation GCT,GTT A636V NP_061170.2
XM_017009621.1 2428 Missense Mutation GCT,GTT A653V XP_016865110.1
XM_017009622.1 2428 Missense Mutation GCT,GTT A641V XP_016865111.1
XM_017009623.1 2428 Intron XP_016865112.1

View Full Product Details