Product Details

SNP ID
rs201297801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179806557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCACAGGCAGAAGAGCAGCAGCGT[C/T]AGGAAGGTGCCATTGCGGAGCCTCA
Phenotype
MIM: 604561 MIM: 601530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGAT4B PubMed Links

Gene Details

Gene
MGAT4B
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014275.4 222 Silent Mutation CTA,CTG L9L NP_055090.1
NM_054013.3 222 Intron NP_463459.1
XM_017008998.1 222 Silent Mutation CTA,CTG L9L XP_016864487.1
Gene
MIR1229
Gene Name
microRNA 1229
There are no transcripts associated with this gene.

Gene
SQSTM1
Gene Name
sequestosome 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142298.1 222 UTR 5 NP_001135770.1
NM_001142299.1 222 Intron NP_001135771.1
NM_003900.4 222 Intron NP_003891.1
XM_011534683.2 222 Intron XP_011532985.1
XM_017010010.1 222 Intron XP_016865499.1

View Full Product Details