Product Details

SNP ID
rs201514329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160393995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCATTAGGATTTGGCCACTAGCT[C/T]GTAATTCAGCAAGATCATTTTTCAT
Phenotype
MIM: 605974 MIM: 615253
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLU7 PubMed Links

Gene Details

Gene
SLU7
Gene Name
SLU7 homolog, splicing factor
There are no transcripts associated with this gene.

Gene
ZBED8
Gene Name
zinc finger BED-type containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303251.1 2078 Missense Mutation CAA,CGA Q499R NP_001290180.1
NM_022090.4 2078 Missense Mutation CAA,CGA Q499R NP_071373.2

View Full Product Details