Product Details

SNP ID
rs200407793
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141135613 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGTCTTGGGGTGGGGGAACTGG[C/T]CACTCGGGGCGCGCGAATGCATTAC
Phenotype
MIM: 606330 MIM: 606331
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PCDHB4 PubMed Links

Gene Details

Gene
PCDHB4
Gene Name
protocadherin beta 4
There are no transcripts associated with this gene.

Gene
PCDHB5
Gene Name
protocadherin beta 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015669.4 408 Missense Mutation GCC,GTC A60V NP_056484.2

View Full Product Details