Product Details

SNP ID
rs201021124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139402025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGGCCAGGAGGACCTTGGTTTT[C/G]GCATGGGCAGCTTTCTCTTCCTGCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC18 PubMed Links

Gene Details

Gene
DNAJC18
Gene Name
DnaJ heat shock protein family (Hsp40) member C18
There are no transcripts associated with this gene.

Gene
PROB1
Gene Name
proline rich basic protein 1
There are no transcripts associated with this gene.

Gene
SPATA24
Gene Name
spermatogenesis associated 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194296.1 231 Silent Mutation GCC,GCG A68A NP_919272.1
XM_005271916.4 231 Silent Mutation GCC,GCG A68A XP_005271973.1
XM_011543252.2 231 Silent Mutation GCC,GCG A29A XP_011541554.1
XM_011543253.2 231 Silent Mutation GCC,GCG A29A XP_011541555.1
XM_011543254.2 231 Silent Mutation GCC,GCG A68A XP_011541556.1

View Full Product Details