Product Details

SNP ID
rs200473303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141095448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAACCCTCACAGCGCTAGATGGC[A/G]GGAGTCCACCCAGGTCCGGCACGGC
Phenotype
MIM: 606328 MIM: 606329
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHB2 PubMed Links

Gene Details

Gene
PCDHB2
Gene Name
protocadherin beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018936.3 842 Missense Mutation AGG,GGG R220G NP_061759.1
Gene
PCDHB3
Gene Name
protocadherin beta 3
There are no transcripts associated with this gene.

View Full Product Details