Product Details

SNP ID
rs200667279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:45261938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGGATCATAAATTTGAAGCAA[A/C]TCGTGGCTTTTCTGCGTCTGGGTCT
Phenotype
MIM: 602780
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HCN1 PubMed Links

Gene Details

Gene
HCN1
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021072.3 2681 Missense Mutation GTT,TTT V886F NP_066550.2

View Full Product Details