Product Details
- SNP ID
-
rs200925434
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:141303177 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGAATTCTGGATTTAATACAATCC[A/T]CTGGGAACACGACAAGCCACAGGCA
- Phenotype
-
MIM: 608157
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC25A2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3749780] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC25A2
- Gene Name
- solute carrier family 25 member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031947.3 |
887 |
Missense Mutation |
GAG,GTG |
E230V |
NP_114153.1 |
View Full Product Details