Product Details

SNP ID
rs200789062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69229216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTTTCCTTTTCAGCGTGGAGG[C/T]CCTGAATAACCATGGTGGCTGCTGT
Phenotype
MIM: 601955 MIM: 611996
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK7 PubMed Links

Gene Details

Gene
CDK7
Gene Name
cyclin dependent kinase 7
There are no transcripts associated with this gene.

Gene
MRPS36
Gene Name
mitochondrial ribosomal protein S36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033281.5 387 Silent Mutation GGC,GGT G101G NP_150597.1

View Full Product Details