Product Details

SNP ID
rs201186087
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141094846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAAACAAAGGCAAGTCCTGATAT[C/T]CTTTGTTTTGCTGGGCATAGCTCAG
Phenotype
MIM: 606328 MIM: 606329
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PCDHB2 PubMed Links

Gene Details

Gene
PCDHB2
Gene Name
protocadherin beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018936.3 240 Missense Mutation TCC,TTC S19F NP_061759.1
Gene
PCDHB3
Gene Name
protocadherin beta 3
There are no transcripts associated with this gene.

View Full Product Details