Product Details
- SNP ID
-
rs201385244
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:176529568 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGTCCGGGTTAAAGCGGTACGAC[A/G]GGAGCTGCTCTATGTCTGCTTTGGT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RNF44
PubMed Links
Gene Details
- Gene
- RNF44
- Gene Name
- ring finger protein 44
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014901.4 |
1507 |
Missense Mutation |
CCG,CTG |
P364L |
NP_055716.1 |
XM_005265840.2 |
1507 |
Missense Mutation |
CCG,CTG |
P364L |
XP_005265897.1 |
XM_005265841.4 |
1507 |
Missense Mutation |
CCG,CTG |
P364L |
XP_005265898.1 |
XM_005265842.4 |
1507 |
Missense Mutation |
CCG,CTG |
P283L |
XP_005265899.1 |
XM_005265843.2 |
1507 |
Missense Mutation |
CCG,CTG |
P283L |
XP_005265900.1 |
XM_005265844.3 |
1507 |
Missense Mutation |
CCG,CTG |
P283L |
XP_005265901.1 |
XM_005265845.4 |
1507 |
Missense Mutation |
CCG,CTG |
P283L |
XP_005265902.1 |
XM_006714831.2 |
1507 |
Missense Mutation |
CCG,CTG |
P361L |
XP_006714894.1 |
XM_006714832.2 |
1507 |
Missense Mutation |
CCG,CTG |
P317L |
XP_006714895.1 |
XM_011534466.2 |
1507 |
Missense Mutation |
CCG,CTG |
P364L |
XP_011532768.1 |
XM_011534467.2 |
1507 |
Missense Mutation |
CCG,CTG |
P283L |
XP_011532769.1 |
View Full Product Details