Product Details

SNP ID
rs200890347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115962641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGGGCCCCCTTCCAGCTCAGG[C/T]TTCTATGTGAGCCGCGCAGTGGCCC
Phenotype
MIM: 610046
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LVRN PubMed Links

Gene Details

Gene
LVRN
Gene Name
laeverin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173800.4 202 Silent Mutation GGC,GGT G8G NP_776161.3
XM_011543263.2 202 Silent Mutation GGC,GGT G8G XP_011541565.1
XM_011543264.2 202 Silent Mutation GGC,GGT G8G XP_011541566.1
XM_017009215.1 202 Silent Mutation GGC,GGT G8G XP_016864704.1

View Full Product Details