Product Details

SNP ID
rs201386463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:146114228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGCTCAGAAATGGGGGTCTTCT[C/T]GGTGTACTCCTTTCCCAGGACAGGA
Phenotype
MIM: 151350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LARS PubMed Links

Gene Details

Gene
LARS
Gene Name
leucyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317964.1 3622 Missense Mutation AAG,GAG K1091E NP_001304893.1
NM_001317965.1 3622 Missense Mutation AAG,GAG K1083E NP_001304894.1
NM_016460.3 3622 Missense Mutation AAG,GAG K1110E NP_057544.2
NM_020117.10 3622 Missense Mutation AAG,GAG K1137E NP_064502.9
XM_011537656.2 3622 Missense Mutation AAG,GAG K1083E XP_011535958.1
Gene
PLAC8L1
Gene Name
PLAC8 like 1
There are no transcripts associated with this gene.

View Full Product Details