Product Details

SNP ID
rs201445934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176881160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGACCCATCCTCTGACTGCAGGAA[C/T]GTGACCACACAGCGAGGGGCCAGCT
Phenotype
MIM: 142570 MIM: 607869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HK3 PubMed Links

Gene Details

Gene
HK3
Gene Name
hexokinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002115.2 2777 Silent Mutation NP_002106.2
XM_011534540.2 2777 Intron XP_011532842.1
XM_017009411.1 2777 Silent Mutation XP_016864900.1
Gene
UNC5A
Gene Name
unc-5 netrin receptor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133369.2 2777 Intron NP_588610.2
XM_006714927.1 2777 Intron XP_006714990.1
XM_006714928.1 2777 Intron XP_006714991.1
XM_011534686.1 2777 Intron XP_011532988.1
XM_011534687.2 2777 Intron XP_011532989.1

View Full Product Details