Product Details
- SNP ID
-
rs200991855
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:23521166 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCAAGTACCTCTGGACAGCACTC[A/T]AGACTAAAACTGGGTAAGAAAAAAT
- Phenotype
-
MIM: 609760
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRDM9
PubMed Links
Gene Details
- Gene
- PRDM9
- Gene Name
- PR domain 9
View Full Product Details