Product Details

SNP ID
rs201012228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177135610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCAGATGTAGATTCTGAAATGGA[C/T]CCAGAACAGCCAGTCACAGAGGATG
Phenotype
MIM: 606681
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NSD1 PubMed Links

Gene Details

Gene
NSD1
Gene Name
nuclear receptor binding SET domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022455.4 645 Silent Mutation GAC,GAT D169D NP_071900.2
NM_172349.2 645 Intron NP_758859.1

View Full Product Details