Product Details

SNP ID
rs201508660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:35904586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTCATCATGATGATGAAGTACA[A/C]ATCAGTGTCAATGGATGCGCTCACA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAPSL PubMed Links

Gene Details

Gene
CAPSL
Gene Name
calcyphosine like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042625.1 663 Missense Mutation NP_001036090.1
NM_144647.3 663 Missense Mutation NP_653248.3
XM_006714444.3 663 Missense Mutation XP_006714507.1
XM_006714445.3 663 Intron XP_006714508.1

View Full Product Details