Product Details

SNP ID
rs201613803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:148313389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTTCAGAAGCTGCTAGTCTGT[A/C]TCCAAAAAAAGTAAGTATGTTGAAT
Phenotype
MIM: 613511
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC102546294 PubMed Links

Gene Details

Gene
LOC102546294
Gene Name
uncharacterized LOC102546294
There are no transcripts associated with this gene.

Gene
SPINK7
Gene Name
serine peptidase inhibitor, Kazal type 7 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032566.2 134 Missense Mutation TAT,TCT Y26S NP_115955.1
Gene
SPINK9
Gene Name
serine peptidase inhibitor, Kazal type 9
There are no transcripts associated with this gene.

View Full Product Details