Product Details

SNP ID
rs201589978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159907021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACGAACATCTCAACTGGCAGGAA[C/T]GCCTCAGTCGGGAATGCACATCGGC
Phenotype
MIM: 165180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAS1 PubMed Links

Gene Details

Gene
MAS1
Gene Name
MAS1 proto-oncogene, G protein-coupled receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002377.2 80 Silent Mutation AAC,AAT N22N NP_002368.1

View Full Product Details