Product Details

SNP ID
rs201747288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41149741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACCAGGACCTTAGGAGGTAGAG[A/G]GGGCAATGAGGATGGAGCTGGGAGT
Phenotype
MIM: 605086 MIM: 609714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCY10P1 PubMed Links

Gene Details

Gene
ADCY10P1
Gene Name
adenylate cyclase 10, soluble pseudogene 1
There are no transcripts associated with this gene.

Gene
LOC105375056
Gene Name
uncharacterized LOC105375056
There are no transcripts associated with this gene.

Gene
TREM2
Gene Name
triggering receptor expressed on myeloid cells 2
There are no transcripts associated with this gene.

Gene
TREML1
Gene Name
triggering receptor expressed on myeloid cells like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271807.1 860 UTR 3 NP_001258736.1
NM_001271808.1 860 Missense Mutation CCT,TCT P156S NP_001258737.1
NM_178174.3 860 Missense Mutation CCT,TCT P267S NP_835468.1
XM_017010822.1 860 Missense Mutation CCT,TCT P326S XP_016866311.1
XM_017010823.1 860 Missense Mutation CCT,TCT P267S XP_016866312.1
XM_017010824.1 860 Missense Mutation CCT,TCT P267S XP_016866313.1
XM_017010825.1 860 UTR 3 XP_016866314.1

View Full Product Details