Product Details
- SNP ID
-
rs201591227
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:10398475 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGGCGTTGTTGTCCGTGTGGCTG[G/T]TGGGGTTGTTGCTGAGGTACATTTT
- Phenotype
-
MIM: 107580
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TFAP2A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3734391] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TFAP2A
- Gene Name
- transcription factor AP-2 alpha
View Full Product Details