Product Details
- SNP ID
-
rs201709066
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:158767015 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCCTTGCCTCATGTTCTCGTTGT[G/T]GATGATGTCATTGTGGGTCCTCTTA
- Phenotype
-
MIM: 123900
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EZR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2242318] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EZR
- Gene Name
- ezrin
- Gene
- MIR3918
- Gene Name
- microRNA 3918
There are no transcripts associated with this gene.
- Gene
- SYTL3
- Gene Name
- synaptotagmin like 3
View Full Product Details