Product Details

SNP ID
rs201947121
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27831251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCGTAGAGGGTGCGGCCCTGGC[G/T]CTTGAGCGCGTAGACCACATCCATG
Phenotype
MIM: 602788 MIM: 602801 MIM: 602826 MIM: 602825
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
HIST1H2AK PubMed Links
Additional Information
For this assay, SNP(s) [rs200488] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HIST1H2AK
Gene Name
histone cluster 1, H2ak
There are no transcripts associated with this gene.

Gene
HIST1H2BN
Gene Name
histone cluster 1, H2bn
There are no transcripts associated with this gene.

Gene
HIST1H4J
Gene Name
histone cluster 1, H4j
There are no transcripts associated with this gene.

Gene
HIST1H4K
Gene Name
histone cluster 1, H4k
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003541.2 Intron NP_003532.1

View Full Product Details