Product Details

SNP ID
rs201948840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46223172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACACGGAGGGTGGCAGGCCAGGA[C/T]GCCGAGTTTGGATGATTTTTGGCTT
Phenotype
MIM: 604876
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RCAN2 PubMed Links

Gene Details

Gene
RCAN2
Gene Name
regulator of calcineurin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251973.1 901 Missense Mutation NP_001238902.1
NM_001251974.1 901 Missense Mutation NP_001238903.1
NM_005822.3 901 Missense Mutation NP_005813.2
XM_011514226.1 901 Missense Mutation XP_011512528.1

View Full Product Details