Product Details

SNP ID
rs202046887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:88047931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGCCCCAGGGGCACGGGCTGCT[C/T]CGCCGGGCTGCTGATGACTGTCGGC
Phenotype
MIM: 612739
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPACA1 PubMed Links

Gene Details

Gene
SPACA1
Gene Name
sperm acrosome associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030960.2 143 Missense Mutation TCC,TTC S9F NP_112222.1
XM_011536160.2 143 Intron XP_011534462.1
XM_017011335.1 143 Intron XP_016866824.1

View Full Product Details