Product Details

SNP ID
rs207467466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:138776131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAACTTAAAACAATGGAAGAGC[A/G]GAAAGTGAAGAGGAGGAGTCCTAAG
Phenotype
MIM: 615353
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC28A PubMed Links

Gene Details

Gene
CCDC28A
Gene Name
coiled-coil domain containing 28A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015439.2 436 Missense Mutation CAG,CGG Q94R NP_056254.1
XM_011535728.2 436 Missense Mutation CAG,CGG Q94R XP_011534030.1
Gene
GVQW2
Gene Name
GVQW motif containing 2
There are no transcripts associated with this gene.

View Full Product Details