Product Details

SNP ID
rs201834523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31639222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGTCGTTTTTGTATATCAGAC[C/T]GGAGCTAAAGAGAAAAAGTAAGCAG
Phenotype
MIM: 142590 MIM: 142580
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAG6 PubMed Links

Gene Details

Gene
BAG6
Gene Name
BCL2 associated athanogene 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098534.1 3622 Missense Mutation CAG,CGG Q1097R NP_001092004.1
NM_001199697.1 3622 Missense Mutation CAG,CGG Q874R NP_001186626.1
NM_001199698.1 3622 Missense Mutation CAG,CGG Q1048R NP_001186627.1
NM_004639.3 3622 Missense Mutation CAG,CGG Q1103R NP_004630.3
NM_080702.2 3622 Missense Mutation CAG,CGG Q1097R NP_542433.1
NM_080703.2 3622 Missense Mutation CAG,CGG Q1097R NP_542434.1
XM_011514892.2 3622 Missense Mutation CAG,CGG Q1144R XP_011513194.1
XM_017011275.1 3622 Missense Mutation CAG,CGG Q1153R XP_016866764.1
XM_017011276.1 3622 Missense Mutation CAG,CGG Q1153R XP_016866765.1
XM_017011277.1 3622 Missense Mutation CAG,CGG Q1153R XP_016866766.1
XM_017011278.1 3622 Missense Mutation CAG,CGG Q1153R XP_016866767.1
XM_017011279.1 3622 Missense Mutation CAG,CGG Q1153R XP_016866768.1
XM_017011280.1 3622 Missense Mutation CAG,CGG Q1153R XP_016866769.1
XM_017011281.1 3622 Missense Mutation CAG,CGG Q1153R XP_016866770.1
XM_017011282.1 3622 Missense Mutation CAG,CGG Q1153R XP_016866771.1
XM_017011283.1 3622 Missense Mutation CAG,CGG Q1152R XP_016866772.1
XM_017011284.1 3622 Missense Mutation CAG,CGG Q1142R XP_016866773.1
XM_017011285.1 3622 Missense Mutation CAG,CGG Q1135R XP_016866774.1
XM_017011286.1 3622 Missense Mutation CAG,CGG Q1135R XP_016866775.1
XM_017011287.1 3622 Missense Mutation CAG,CGG Q1133R XP_016866776.1
XM_017011288.1 3622 Missense Mutation CAG,CGG Q1114R XP_016866777.1
XM_017011289.1 3622 Missense Mutation CAG,CGG Q1106R XP_016866778.1
XM_017011290.1 3622 Missense Mutation CAG,CGG Q1105R XP_016866779.1
XM_017011291.1 3622 Missense Mutation CAG,CGG Q1104R XP_016866780.1
XM_017011292.1 3622 Missense Mutation CAG,CGG Q1096R XP_016866781.1
XM_017011293.1 3622 Missense Mutation CAG,CGG Q1079R XP_016866782.1
XM_017011294.1 3622 Missense Mutation CAG,CGG Q1071R XP_016866783.1
XM_017011295.1 3622 Missense Mutation CAG,CGG Q1056R XP_016866784.1
XM_017011296.1 3622 Missense Mutation CAG,CGG Q1047R XP_016866785.1
XM_017011297.1 3622 Missense Mutation CAG,CGG Q1065R XP_016866786.1
XM_017011298.1 3622 Missense Mutation CAG,CGG Q999R XP_016866787.1
Gene
MIR6832
Gene Name
microRNA 6832
There are no transcripts associated with this gene.

Gene
PRRC2A
Gene Name
proline rich coiled-coil 2A
There are no transcripts associated with this gene.

View Full Product Details