Product Details

SNP ID
rs201934364
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52757868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTAACTCAGAACCTACCTCTACT[C/G]CAGCTGCAGCCAGGAGCCACCGGAT
Phenotype
MIM: 138360
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
GSTA2 PubMed Links

Gene Details

Gene
GSTA2
Gene Name
glutathione S-transferase alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000846.4 230 Missense Mutation GCA,GGA A27G NP_000837.3
XM_011514532.2 230 Intron XP_011512834.1

View Full Product Details