Product Details

SNP ID
rs201738462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35805870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGCTACTTCGGCCTTTTCTCCT[A/G]CTGCGTGGGTAACGTGCTGTCCTCC
Phenotype
MIM: 120105 MIM: 609427
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLPS PubMed Links

Gene Details

Gene
CLPS
Gene Name
colipase
There are no transcripts associated with this gene.

Gene
LHFPL5
Gene Name
lipoma HMGIC fusion partner-like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182548.3 577 Missense Mutation TAC,TGC Y67C NP_872354.1

View Full Product Details