Product Details

SNP ID
rs201996097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116877498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCTGCCGGGGGCAGTGAAATCA[C/G]GTGAGTGCTCTGCCGCATCCGGAGC
Phenotype
MIM: 612659
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RFX6 PubMed Links

Gene Details

Gene
RFX6
Gene Name
regulatory factor X6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173560.3 286 Missense Mutation CAA,GAA Q75E NP_775831.2
XM_011535589.1 286 Missense Mutation CAA,GAA Q75E XP_011533891.1
XM_017010477.1 286 Intron XP_016865966.1

View Full Product Details