Product Details

SNP ID
rs201624028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41149812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAGAGGTAGGCTGGTGTAGGTG[A/G]TATTGTCAAATGAAGGTGGTGAGTC
Phenotype
MIM: 605086 MIM: 609714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCY10P1 PubMed Links

Gene Details

Gene
ADCY10P1
Gene Name
adenylate cyclase 10, soluble pseudogene 1
There are no transcripts associated with this gene.

Gene
LOC105375056
Gene Name
uncharacterized LOC105375056
There are no transcripts associated with this gene.

Gene
TREM2
Gene Name
triggering receptor expressed on myeloid cells 2
There are no transcripts associated with this gene.

Gene
TREML1
Gene Name
triggering receptor expressed on myeloid cells like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271807.1 789 UTR 3 NP_001258736.1
NM_001271808.1 789 Missense Mutation ACC,ATC T132I NP_001258737.1
NM_178174.3 789 Missense Mutation ACC,ATC T243I NP_835468.1
XM_017010822.1 789 Missense Mutation ACC,ATC T302I XP_016866311.1
XM_017010823.1 789 Missense Mutation ACC,ATC T243I XP_016866312.1
XM_017010824.1 789 Missense Mutation ACC,ATC T243I XP_016866313.1
XM_017010825.1 789 UTR 3 XP_016866314.1

View Full Product Details